Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRB2-related adaptor protein 2, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018164 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-164 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-164 Supplier Abnova Corporation Supplier No. H00009402D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human GRAP2 protein.

This gene encodes a member of the GRB2/Sem5/Drk family. This member is an adaptor-like protein involved in leukocyte-specific protein-tyrosine kinase signaling. Like its related family member, GRB2-related adaptor protein (GRAP), this protein contains an SH2 domain flanked by two SH3 domains. This protein interacts with other proteins, such as GRB2-associated binding protein 1 (GAB1) and the SLP-76 leukocyte protein (LCP2), through its SH3 domains. Transcript variants utilizing alternative polyA sites exist. [provided by RefSeq

Sequence: MEAVAKFDFTASGEDELSFHTGDVLKILSNQEEWFKAELGSQEGYVPKNFIDIQFPKWFHEGLSRHQAENLLMGKEVGFFIIRASQSSPGDFSISVRHEDDVQHFKVMRDNKGNYFLWTEKFPSLNKLVDYYRTNSISRQKQIFLRDRTREDQGHRGNSLDRRSQGGPHLSGAVGEEIRPSMNRKLSDHPPTLPLQQHQHQPQPPQYAPAPQQLQQPPQQRYLQHHHFHQERRGGSLDINDGHCGTGLGSEMNAALMHRRHTDPVQLQAAGRVRWARALYDFEALEDDELGFHSGEVVEVLDSSNPSWWTGRLHNKLGLFPANYVAPMTR

Specifications

Antigen GRB2-related adaptor protein 2
Applications In situ PLA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human GRAP2 protein.
Formulation PBS with no preservative; pH 7.4
Gene GRAP2
Gene Accession No. NM_004810
Gene Alias GADS/GRAP-2/GRB2L/GRBLG/GRID/GRPL/GrbX/Grf40/Mona/P38
Gene Symbols GRAP2
Host Species Rabbit
Immunogen GRAP2 (NP_004801.1, 1 a.a. ∼ 330 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Membrane Receptors
Primary or Secondary Primary
Gene ID (Entrez) 9402
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.