Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRB2 Mouse, Unlabeled, Clone: 81, BD

Catalog No. BDB610111 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610111 50 μg
BDB610112 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610111 Supplier BD Biosciences Supplier No. 610111
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Growth-factor Receptor-Bound Protein 2 (GRB2) was isolated by using the EGF receptor C-terminus as a probe in the screening of λgt11 expression libraries. This 24kDa GRB2 protein is ubiquitously expressed and consists of one SH2 domain flanked by two SH3 domains. Its ability to bind proteins through these domains has prompted much investigation of its role as an adaptor protein. Sos, a Ras GDP/GTP exchange protein, is constitutively bound to the SH3 domain of GRB2. Following growth factor stimulation, receptor tyrosine kinases autophosphorylate, creating a binding site for the GRB2 SH2 domain. Alternatively, GRB2 interacts with the receptor-bound active Shc protein.Through these interactions, the GRB2/Sos complex is translocated to the membrane where Sos activates membrane-bound Ras to initiate the Ras signaling pathway. Other proteins that similarly interact with GRB2 include Cbl, PTP1D, and Dynamin. However,the mechanisms through which these associations impact cellular responses remain to be discovered.

Host Species: Mouse
Clone: 81
Isotype: IgG1
Species Reactivity: Rat
Immunogen: Rat GRB2 aa. 1-217
Formula Weight [Chemical]: 24kDa

Bioimaging, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen GRB2
Applications Western Blot
Classification Monoclonal
Clone 81
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Rat GRB2 aa. 1-217
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Frog, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.