Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Green Fluorescent Protein Mouse anti-All, Clone: 264-449-2, MilliporeSigma™

Catalog No. MAB2510MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2510MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB2510MI Supplier MilliporeSigma Supplier No. MAB2510
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Green Fluorescent Protein Antibody, original GFP only, clone 264-449-2 is an antibody against Green Fluorescent Protein for use in immunoprecipitation, western blotting and immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Green Fluorescent Protein
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 264-449-2
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid in 0.02M Phosphate buffer, 0.25M NaCl, pH 7.6 with 0.1% sodium azide.
Host Species Mouse
Immunogen highly-purified native GFP
Quantity 100 μg
Primary or Secondary Primary
Target Species All species
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.