Learn More
Green Fluorescent Protein Mouse anti-All, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB3580
Description
Detect Green Fluorescent Protein using this Anti-Green Fluorescent Protein Antibody validated for use in ELISA, immunocytochemistry, immunohistochemistry and western blotting.
Specifications
Green Fluorescent Protein | |
Monoclonal | |
Liquid in PBS containing 1mg/mL BSA. Contains no preservative. | |
GFP-BSA | |
100 μL | |
All species | |
Purified |
ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot | |
Unconjugated | |
Mouse | |
Protein A purfied | |
Primary | |
−20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. | |
IgG1 |