Learn More
MilliporeSigma™ anti-Green Fluorescent Protein (GFP; eGFP); Polyclonal; ™ (Chemicon™)
Specifications
| Antigen | Green Fluorescent Protein (GFP; eGFP) |
|---|---|
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Semi-purified |
| Host Species | Rabbit |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
50-173-418
|
MilliporeSigma™
AB3080/DEL |
50 μg | N/A | N/A | N/A | ||||
Description
The Green Fluorescent Protein (GFP) from the jellyfish Aequorea victoria is used as a fluorescent indicator for monitoring gene expression in a variety of cellular systems, including living organisms and fixed tissues. Unlike other bioluminescent reporters, GFP fluoresces in the absence of substrates, cofactors, or other intrinsic or extrinsic proteins. Purified GFP is a 27kDa monomer consisting of 238 amino acids and emits green light (emission maximum at 509nm) when excited with blue or UV light.
ELISA, Immunocytochemistry, Immunohistochemistry, Western Blotting
Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly
Specifications
| Green Fluorescent Protein (GFP; eGFP) | |
| Unconjugated | |
| Rabbit | |
| Epitope Tags | |
| Highly purified native GFP from AHuorea victoria. |
| Polyclonal | |
| Semi-purified | |
| RUO | |
| Vertebrates | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.