Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GREM1, Mouse, Clone: 2C8, Abnova™

Catalog No. 89123129 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-123-129 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-123-129 Supplier Abnova Corporation Supplier No. H00026585M02A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant GREM1.

This gene encodes a member of the BMP (bone morphogenic protein) antagonist family. Like BMPs, BMP antagonists contain cystine knots and typically form homo- and heterodimers. The CAN (cerberus and dan) subfamily of BMP antagonists, to which this gene belongs, is characterized by a C-terminal cystine knot with an eight-membered ring. The antagonistic effect of the secreted glycosylated protein encoded by this gene is likely due to its direct binding to BMP proteins. As an antagonist of BMP, this gene may play a role in regulating organogenesis, body patterning, and tissue differentiation. In mouse, this protein has been shown to relay the sonic hedgehog (SHH) signal from the polarizing region to the apical ectodermal ridge during limb bud outgrowth. [provided by RefSeq

Sequence: ESSQEALHVTERKYLKRDWCKTQPLKQTIHEEGCNSRTIINRFCYGQCNSFYIPRHIRKEEGSFQSCSFCKPKKFTTMMVTLNCPELQPPTKKKRVTRVKQCRCISIDLD

Specifications

Antigen GREM1
Applications ELISA
Classification Monoclonal
Clone 2C8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant GREM1.
Formulation ascites with no preservative
Gene GREM1
Gene Accession No. NM_013372
Gene Alias CKTSF1B1/DAND2/DRM/GREMLIN/IHG-2/MGC126660/PIG2
Gene Symbols GREM1
Host Species Mouse
Immunogen GREM1 (NP_037504, 75 a.a. ∼ 184 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26585
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.