Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

glutamate receptor, ionotrophic, AMPA 3, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89005472 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-005-472 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-005-472 Supplier Abnova Corporation Supplier No. H00002892B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human GRIA3 protein.

Glutamate receptors are the predominant excitatory neurotransmitter receptors in the mammalian brain and are activated in a variety of normal neurophysiologic processes. These receptors are heteromeric protein complexes composed of multiple subunits, arranged to form ligand-gated ion channels. The classification of glutamate receptors is based on their activation by different pharmacologic agonists. The subunit encoded by this gene belongs to a family of AMPA (alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate)-sensitive glutamate receptors, and is subject to RNA editing (AGA->GGA; R->G). Alternative splicing at this locus results in different isoforms, which may vary in their signal transduction properties. [provided by RefSeq

Sequence: MARQKKMGQSVLRAVFFLVLGLLGHSHGGFPNTISIGGLFMRNTVQEHSAFRFAVQLYNTNQNTTEKPFHLNYHVDHLDSSNSFSVTNACPAERDYLPWPGSIRENNWTALPCCKDHGLLHLKCSPGGARQNWAYCIWGVTGEL

Specifications

Antigen glutamate receptor, ionotrophic, AMPA 3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human GRIA3 protein.
Formulation No additive
Gene GRIA3
Gene Accession No. NM_181894
Gene Alias GLUR-C/GLUR-K3/GLUR3/GLURC/MRX94
Gene Symbols GRIA3
Host Species Mouse
Immunogen GRIA3 (NP_871623, 1 a.a. ∼ 144 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 2892
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.