Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRID2, Mouse, Clone: 1A1, Abnova™

Catalog No. 89022386 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-022-386 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-022-386 Supplier Abnova Corporation Supplier No. H00002895M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant GRID2.

Human glutamate receptor delta-2 (GRID2) is a relatively new member of the family of ionotropic glutamate receptors which are the predominant excitatory neurotransmitter receptors in the mammalian brain. GRID2 is a predicted 1007 amino acid protein that shares 97% identity with the mouse homolog which is expressed selectively in cerebellar Purkinje cells. A point mutation in mouse GRID2, associated with the phenotype named 'lurcher', in the heterozygous state leads to ataxia resulting from selective, cell-autonomous apoptosis of cerebellar Purkinje cells during postnatal development. Mice homozygous for this mutation die shortly after birth from massive loss of mid- and hindbrain neurons during late embryogenesis. This strongly suggests a role for GRID2 in neuronal apoptotic death. [provided by RefSeq]

Sequence: DTLPTRQALEQISDFRNTHITTTTFIPEQIQTLSRTLSAKAASGFTFGNVPEHRTGPFRHRAPNGGFFRSPIKTMSSIPYQPTPTLGLNLGNDPDRGTSI

Specifications

Antigen GRID2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1A1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant GRID2.
Formulation PBS with no preservative; pH 7.4
Gene GRID2
Gene Accession No. NM_001510
Gene Alias MGC117022/MGC117023/MGC117024
Gene Symbols GRID2
Host Species Mouse
Immunogen GRID2 (NP_001501, 908 a.a. ∼ 1007 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2895
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.