Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRIK3, Mouse, Clone: 4H1, Abnova™

Catalog No. 89022389 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-022-389 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-022-389 Supplier Abnova Corporation Supplier No. H00002899M06
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant GRIK3.

Glutamate receptors are the predominant excitatory neurotransmitter receptors in the mammalian brain and are activated in a variety of normal neurophysiologic processes. This gene product belongs to the kainate family of glutamate receptors, which are composed of four subunits and function as ligand-activated ion channels. It is not certain if the subunit encoded by this gene is subject to RNA editing as the other 2 family members (GRIK1 and GRIK2). A Ser310Ala polymorphism has been associated with schizophrenia, and there are conflicting reports of its association with the pathogenesis of delirium tremens in alcoholics. [provided by RefSeq

Sequence: NLYPDYASLSHAILDLVQYLKWRSATVVYDDSTGLIRLQELIMAPSRYNIRLKIRQLPIDSDDSRPLLKEMKRGREFRIIFDCSHTMAAQILKQAMAMGM

Specifications

Antigen GRIK3
Applications ELISA
Classification Monoclonal
Clone 4H1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant GRIK3.
Formulation PBS with no preservative; pH 7.4
Gene GRIK3
Gene Accession No. NM_000831
Gene Alias EAA5/GLR7/GLUR7/GluR7a
Gene Symbols GRIK3
Host Species Mouse
Immunogen GRIK3 (NP_000822, 151 a.a. ∼ 250 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2899
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.