Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

glutamate receptor, ionotropic, N-methyl D-aspartate 2C, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89005474 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-005-474 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-005-474 Supplier Abnova Corporation Supplier No. H00002905B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human GRIN2C protein.

N-methyl-D-aspartate (NMDA) receptors are a class of ionotropic glutamate receptors. NMDA channel has been shown to be involved in long-term potentiation, an activity-dependent increase in the efficiency of synaptic transmission thought to underlie certain kinds of memory and learning. NMDA receptor channels are heteromers composed of the key receptor subunit NMDAR1 (GRIN1) and 1 or more of the 4 NMDAR2 subunits: NMDAR2A (GRIN2A), NMDAR2B (GRIN2B), NMDAR2C (GRIN2C), and NMDAR2D (GRIN2D).

Sequence: MGGALGPALLLTSLFGAWAGLGPGQGEQGMTVAVVFSSSGPPQAQFRARLTPQSFLDLPLEIQPLTVGVNTTNPSSLLTQICGLLGAAHVHGIVFEDNVDTEAVAQILDFISSQTHVPILSISGGSAVVLTPKVHVQTHVPSCLRPGTRLGSGVLWFWEAGIRRDGQGGGG

Specifications

Antigen glutamate receptor, ionotropic, N-methyl D-aspartate 2C
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human GRIN2C protein.
Formulation No additive
Gene GRIN2C
Gene Accession No. BC059384
Gene Alias NMDAR2C/NR2C
Gene Symbols GRIN2C
Host Species Mouse
Immunogen GRIN2C (AAH59384, 1 a.a. ∼ 171 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 2905
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.