Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRINL1A, Rabbit, Monoclonal Antibody, Abnova™

Catalog No. 89203752 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg x up to 3 Abs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-203-752 1 mg x up to 3 Abs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-203-752 Supplier Abnova Corporation Supplier No. H00081488K

Includes

Up to three rabbit IgG clones of 1mg each will be delivered to customer.

Rabbit monoclonal antibody raised against a human GRINL1A peptide using ARM Technology.

This gene (GRINL1A) is part of a complex transcript unit that includes the gene for GRINL1A combined protein (Gcom1). Transcription of this gene occurs at a downstream promoter, with at least three different alternatively spliced variants, grouped together as Gdown for GRINL1A downstream transcripts. The Gcom1 gene uses an upstream promoter for transcription and also has multiple alternatively spliced variants. [provided by RefSeq

Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)
Expression: Overexpression vector and transfection into 293H cell line

Order Info

  1. Customer may provide cell or tissue lysate for antibody screening.
  2. Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.

Specifications

Antigen GRINL1A
Applications ELISA, Western Blot
Classification Monoclonal
Concentration 1mg
Conjugate Unconjugated
Description glutamate receptor, ionotropic, N-methyl D-aspartate-like 1A
Formulation In 1X PBS, pH 7.4
Gene GRINL1A
Gene Accession No. GRINL1A
Gene Alias DKFZp586F1918
Gene Symbols GRINL1A
Host Species Rabbit
Immunogen A synthetic peptide of human GRINL1A
Quantity 1 mg x up to 3 Abs
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81488
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.