Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GRIP1 Polyclonal Antibody

GRIP1 Polyclonal Antibody
Supplier: Thermo Scientific PA534510
Description
GRIP1 Polyclonal Antibody for Western Blot, IHC
Specifications
| GRIP1 | |
| Polyclonal | |
| Unconjugated | |
| GRIP1 | |
| 4931400F03Rik; AMPA receptor-interacting protein GRIP1; eb; eye blebs; glutamate receptor interacting protein 1; glutamate receptor-interacting protein 1; GRIP; Grip1; GRIP-1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23426, 74053, 84016 | |
| Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| P97879, Q925T6, Q9Y3R0 | |
| GRIP1 | |
| A 17 amino acid peptide near the carboxy terminus of human GRIP1. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction