Learn More
GRIPAP1, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human GRIPAP1 peptide using ARM Technology.
Supplier: Abnova Corporation H00056850K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
GRASP1 is a neuron-specific guanine nucleotide exchange factor for the Ras family of small G proteins (RasGEF) and is associated with the GRIP/AMPA receptor complex in brain (Ye et al., 2000 [PubMed 10896157]).[supplied by OMIM
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
GRIPAP1 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
GRIPAP1 | |
GRIPAP1 | |
A synthetic peptide of human GRIPAP1 | |
RUO | |
56850 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
GRIP1 associated protein 1 | |
GRIPAP1 | |
DKFZp434P0630, GRASP-1, KIAA1167, MGC126593, MGC126595 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.