Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRK 4-6 Mouse anti-Human, Mouse, Rat, Clone: A16/17, MilliporeSigma™

Catalog No. 05466MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-466-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-466-MI Supplier MilliporeSigma Supplier No. 05466
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-GRK 4-6 Antibody, clone A16/17 detects level of GRK 4-6 and has been published and validated for use in immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GRK 4-6
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone A16/17
Conjugate Unconjugated
Formulation 0.1M Tris-glycine, pH 7.4, 0.15 NaCl with 0.05% sodium azide
Gene Accession No. NM_019497
Gene Symbols GRK4, GRK4a, IT11, GPRK4, GPRK2L, ITI1
Host Species Mouse
Immunogen GST fusion protein corresponding to residues 463-590 of bovine GRK 5
Purification Method Ammonium sulfate precipitation
Quantity 200 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage −20°C for 24 months.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.