Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GRLF1, Mouse, Clone: 4D4-F8, Abnova™

Catalog No. 89022392 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-022-392 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-022-392 Supplier Abnova Corporation Supplier No. H00002909M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant GRLF1.

The human glucocorticoid receptor DNA binding factor, which associates with the promoter region of the glucocorticoid receptor gene (hGR gene), is a repressor of glucocorticoid receptor transcription. The amino acid sequence deduced from the cDNA sequences show the presence of three sequence motifs characteristic of a zinc finger and one motif suggestive of a leucine zipper in which 1 cysteine is found instead of all leucines. The GRLF1 enhances the homologous down-regulation of wild-type hGR gene expression. Biochemical analysis suggests that GRLF1 interaction is sequence specific and that transcriptional efficacy of GRLF1 is regulated through its interaction with specific sequence motif. The level of expression is regulated by glucocorticoids. [provided by RefSeq

Sequence: MEATSRSVHGDVGEVGHFEGMAVCWCPRRGILPGLR

Specifications

Antigen GRLF1
Applications ELISA
Classification Monoclonal
Clone 4D4-F8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant GRLF1.
Formulation PBS with no preservative; pH 7.4
Gene GRLF1
Gene Accession No. BC003514
Gene Alias GRF-1/KIAA1722/MGC10745/P190-A/P190A/p190RhoGAP
Gene Symbols GRLF1
Host Species Mouse
Immunogen GRLF1 (AAH03514, 1 a.a. ∼ 36 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2909
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.