Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Growth Associated Protein 43 Rabbit anti-Bovine, Human, Mouse, Monkey, Rat, Feline, Polyclonal, MilliporeSigma™

Catalog No. AB5312MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB5312MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB5312MI Supplier MilliporeSigma Supplier No. AB5312

Rabbit Polyclonal Antibody

This Anti-Growth Associated Protein 43 Antibody is validated for use in immunocytochemistry, immunohistochemistry, immunohistochemistry (paraffin), immunoprecipitation, western blotting for the detection of Growth Associated Protein 43.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Growth Associated Protein 43
Applications Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Serum. Liquid with 0.02% sodium azide added as a preservative.
Gene Symbols GAP43; PP46; Neuromodulin; B-50; neuromodulin
Host Species Rabbit
Immunogen Purified GAP-43 from feline brain (McIntosh and Parkinson, 1990).
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_002045.2
Target Species Bovine, Feline, Human, Monkey, Mouse, Rat
Content And Storage −20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.