Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Growth Associated Protein-43 (GAP-43) Rabbit anti-Human, Mouse, Rat, Chicken, Bovine, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB5220
Description
Anti-Growth Associated Protein-43 (GAP-43) Antibody is an antibody against Growth Associated Protein-43 (GAP-43) for use in ELISA, immunocytochemistry, immunohistochemistry, immunoprecipitation and western blotting.
Specifications
Growth Associated Protein 43 | |
Polyclonal | |
Purified rabbit polyclonal in buffer containing PBS with 0.01% sodium azide as a preservative. | |
Rabbit | |
Ammonium sulfate precipitation | |
Primary | |
Bovine, Chicken, Human, Mouse, Rat | |
Purified |
ELISA, Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
Unconjugated | |
GAP43; PP46; B-50 | |
Recombinant rat GAP-43 (complete sequence). | |
100 μL | |
NM_002045.2 | |
-20°C in undiluted aliquotes for up to 6 months, Avoid Freeze/Thaw Cycles |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction