Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Growth Associated Protein 43 Mouse anti-Human, Rat, Feline, Hamster, Clone: 9-1E12, MilliporeSigma™

Catalog No. MAB347MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB347MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB347MI Supplier MilliporeSigma Supplier No. MAB347
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Growth Associated Protein 43 Antibody, clone 9-1E12 detects level of Growth Associated Protein 43 and has been published and validated for use in immunohistochemistry, immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Growth Associated Protein 43
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 9-1E12
Conjugate Unconjugated
Formulation Protein A Purified mouse immunoglobulin IgG1 in 20mM sodium phosphate, 250mM NaCl, pH. 7.6, with 0.1% sodium azide as a preservative.
Gene Accession No. NM_002045.2
Gene Symbols GAP43, PP46, B-50, GAP-43/B50
Host Species Mouse
Immunogen GAP-43 purified from rat brain
Purification Method Protein A purfied
Quantity 50 μg
Primary or Secondary Primary
Target Species Feline, Hamster, Human, Rat
Content And Storage 2-8°C in undiluted aliquots for up to 6 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.