Learn More
Growth Associated Protein 43 Rabbit anti-Rat, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Growth Associated Protein 43, phosphoSer 41 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity purified immunoglobulin. Liquid in 10mM HEPES (pH 7.5), 150mM NaCl with 100mg/mL BSA and 50% glycerol. |
| Gene Symbols | GAP43; PP46; Neuromodulin; B-50; neuromodulin |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide corresponding to amino acids surrounding the phosphoSer41 of rat GAP-43. This is the site phosphorylated by protein kinase C. |
| Purification Method | Affinity Purified |
| Quantity | 25 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.