Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gsα Rabbit anti-Bovine, Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. 06237MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-237-MI 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-237-MI Supplier MilliporeSigma Supplier No. 06237
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Gsα Antibody is an antibody against Gsα for use in immunocytochemistry, immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Gsα
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Serum diluted 1:1 in PBS.
Gene Symbols GNAS; NESP55; GNASXL; C20orf45; GPSA; dJ309F20.1.1; GSA; PHP1B; CARD3; SCG6; MGC33735; GSP; dJ806M20.3.3; PHP1A; GNAS1; POH; AHO; NESP; XLalphas
Host Species Rabbit
Immunogen peptide corresponding to the c-terminus of bovine Gsα (RMHLRQYELL); this sequence is present in all three isoforms of human Gsα (Gsα1, Gsα2, and GsαXL)
Purification Method Protein A purfied
Quantity 50 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_000516.4; NM_001077488.1; NM_001077489.1; NM_001077490.1; NM_016592.2; NM_080425.2; NM_080426.2
Target Species Bovine, Human, Mouse
Content And Storage −20°C in undiluted aliquotes for 24 months. Avoid repeated freeze/thaw cycles.
Form Serum
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.