Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSAP Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP178376 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP178376 0.1 mL
NBP178376SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP178376 Supplier Novus Biologicals Supplier No. NBP178376
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GSAP Polyclonal specifically detects GSAP in Human samples. It is validated for Western Blot, Simple Western.

Specifications

Antigen GSAP
Applications Western Blot
Classification Polyclonal
Concentration 1.11 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Simple Western 1:100
Formulation PBS and 30% Glycerol with 0.05% Sodium Azide
Gene Accession No. A4D1B5
Gene Alias Gamma-secretase-activating protein, pigeon homolog (Drosophila)
Gene Symbols PION
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human GSAP protein (between residues 100-150) [UniProt A4D1B5]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Neurodegeneration, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 54103
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.