Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSK-3b Rabbit anti-Rat, Human, Mouse, Bovine, Polyclonal, MilliporeSigma™

Catalog No. 071413MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-141-3MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-141-3MI Supplier MilliporeSigma Supplier No. 071413
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This AntiI-GSK-3b Antibody, a.a. 335-349 is validated for use in western blot, immunoprecipitation, immunohistochemistry (paraffin) for the detection of AntiI-GSK-3b.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GSK-3b
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified polyclonal IgG antibody is supplied in PBS, pH 7.2, 50% glycerol and 0.09% sodium azide.
Gene Symbols GSK3B; EC 2.7.11.26
Host Species Rabbit
Immunogen Rat GSK3 beta synthetic peptide (aa. 335-349), conjugated to KLH. This sequence is 100% identical to human GSK3 beta.
Purification Method Affinity Purified
Quantity 50 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_002093
Target Species Bovine, Human, Mouse, Rat
Content And Storage −20°C for up to 12 months
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.