Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-GSK-3beta, Polyclonal

Catalog No. PK1111100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
PK1111100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PK1111100UG Supplier MilliporeSigma™ Supplier No. PK1111100UG

Rabbit Polyclonal Antibody

Specifically detects GSK-3β Clone: in Human, Mouse, Rat samples, and it is validated for Immunohistochemistry

Glycogen synthase kinase 3 (GSK-3) is a serine/threonine protein kinase that plays a role in the regulation of cell survival and in the Wnt signaling pathway. Its activity can be inhibited by Akt-mediated phosphorylation at Ser21 in GSK-3α and Ser9 in GSK-3β. GSK-3 is also known to phosphorylate the six isoforms of Tau, indicating a link to Alzheimer's disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GSK-3β
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation In PBS, 30% glycerol, 0.5% BSA.
Host Species Rabbit
Immunogen recombinant human GSK-3β
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽46kDa GSK-3β protein in mouse small intestine tissue.
Target Species Human, Mouse, Rat
Content And Storage −20°C
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.