Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSK3 Mouse anti-Vertebrates, Mouse, Rat, Human, Dictyostelium, Yeast (S. pombe), Clone: 4G-1E, MilliporeSigma™

Catalog No. 05412MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-412-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-412-MI Supplier MilliporeSigma Supplier No. 05412
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-GSK3 Antibody, clone 4G-1E detects level of GSK3 and has been published and validated for use in IC and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GSK3
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 4G-1E
Conjugate Unconjugated
Formulation Protein G purified mouse IgG1 in storage buffer containing 0.02Mphosphate buffer, pH 7.6, 0.25MNaCl, and 0.1% sodium azide.
Gene Accession No. NM_057366
Gene Symbols GSK3A, DKFZp686D0638
Host Species Mouse
Immunogen 18 residue synthetic peptide (C-KQLLHGEP NVSYICSRY) corresponding to amino acids 203-219 derived from a region of the catalytic domain (between subdomain VII and VIII) of the Drosophila GSK3/shaggy enzyme, coupled to ovalbumin. Clone 4G-1E.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species D. discoideum, Human, Mouse, Rat, Vertebrates, Yeast
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.