Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSK3 Mouse anti-Human, FITC, Clone: 4G-1E, MilliporeSigma™

Catalog No. FCMAB386FMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FCMAB386FMI 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FCMAB386FMI Supplier MilliporeSigma Supplier No. FCMAB386F

Mouse Monoclonal Antibody

Milli-Mark™ Anti-GSK3-FITC Antibody, clone 4G-1E detects level of GSK3 and has been published and validated for use in flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GSK3
Applications Flow Cytometry
Classification Monoclonal
Clone 4G-1E
Conjugate FITC
Formulation Purified mouse monoclonal IgG1 conjugated to FITC in PBS with 0.1% sodium azide and 15mg/mL BSA.
Gene Accession No. NP_063937
Gene Symbols GSK3A, DKFZp686D0638
Host Species Mouse
Immunogen Synthetic peptide derived from a region of the catalytic domain (between subdomain VII and VIII) of the Drosophila GSK3/shaggy enzyme, coupled to ovalbumin.
Purification Method Protein G Purified
Quantity 100 Tests
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 6 months. Protect from Light.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.