Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GSK3α/Β Rabbit anti-Human, Mouse, Rat, MilliporeSigma™
Rabbit Monoclonal Antibody
Supplier: MilliporeSigma 04903
Description
This Anti-GSK3α/β Antibody is validated for use in western blot, immunoprecipitation for the detection of GSK3α/β.
Specifications
GSK3α/β | |
Monoclonal | |
Cultured supernantant containing 0.05% sodium azide. | |
Rabbit | |
100 μL | |
DKFZp686D0638, EC 2.7.11.26 | |
−20°C for 12 months. Aliquot to avoid freeze/thaw cycles. | |
IgG |
Immunoprecipitation, Western Blot | |
Unconjugated | |
NP_063937.2, NP_002084.2 | |
N-terminal 6His-tagged recombinant full length human GSK3β. | |
Primary | |
Human, Mouse, Rat | |
Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction