Learn More
Description
The protein encoded by this gene is a serine-threonine kinase, belonging to the glycogen synthase kinase subfamily. It is involved in energy metabolism, neuronal cell development, and body pattern formation. Polymorphisms in this gene have been implicated in modifying risk of Parkinson disease, and studies in mice show that overexpression of this gene may be relevant to the pathogenesis of Alzheimer disease. Alternatively spliced transcript variants encoding different isoforms have been found for this gene
Specifications
Specifications
| Antigen | glycogen synthase kinase 3 beta |
| Applications | ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), In situ PLA, Western Blot |
| Classification | Monoclonal |
| Clone | 1H3 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against a partial recombinant GSK3B. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | GSK3B |
| Gene Accession No. | NM_002093 |
| Gene Symbols | GSK3B |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.