Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-GST, Polyclonal, Cytiva

Catalog No. 45001369 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
500 μL (50 Detections)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-001-369 500 μL (50 Detections)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-001-369 Supplier Cytiva Supplier No. 27457701
Only null left
Add to Cart
Add to Cart

Detect recombinant schistosomal GST fusion proteins expressed using pGEX vectors. Anti-GST, Polyclonal, Cytiva, is optimized for sensitive and specific applications.

Host Species: Goat
Immunogen: Purified schistosomal GST

Dot Blotting, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen GST
Conjugate Unconjugated
Host Species Goat
Immunogen Purified schistosomal GST
Purification Method Affinity Purified
Quantity 500 μL (50 Detections)
Primary or Secondary Primary
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.