Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GST Tag Mouse anti-Schistosoma japonicum, HRP, Clone: DG122-2A7, MilliporeSigma™

Catalog No. 16209MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-209-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 16-209-MI Supplier MilliporeSigma Supplier No. 16209
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-GST Tag Antibody, clone DG122-2A7, HRP conjugate detects level of GST Tag and has been published and validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GST Tag
Applications Western Blot
Classification Monoclonal
Clone DG122-2A7
Conjugate HRP
Formulation Purified mouse monoclonal IgG in buffer containing (HRP) in 70% storage buffer PBS with 0.05% Kathon™ and 30% glycerol. Liquid at −20°C.
Gene Accession No. NP_001070
Gene Symbols ZAP70, ZAP-70, TZK, STD, SRK
Host Species Mouse
Immunogen GST (Glutathione S-transferase) derived from the pGEX expression vector and linked to KLH. Clone DG122-2A7.
Purification Method Protein G purfied
Quantity 100 μL
Primary or Secondary Primary
Target Species S. japonicum
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.