Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-GSTA4, Polyclonal

Catalog No. ABS1652MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABS1652MI 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS1652MI Supplier MilliporeSigma™ Supplier No. ABS1652
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects GSTA4 Clone: in Mouse, Rat samples, and it is validated for Immunohistochemistry (Paraffin), Western Blotting

Glutathione S-transferase alpha-4 (EC:2.5.1.18; UniProt P14942; also known as Glutathione S-transferase A4, Glutathione S-transferase Yk, GST 8-8, GST A4-4, GST K, GST Yk) is encoded by the Gsta4 gene (Gene ID 300850) in rat. Glutathione S-transferases constitute a family of isozymes that are involved in detoxification by catalyzing the conjugation of the reduced form of glutathione (GSH) to xenobiotic substrates. The GST family can be subdivided into the cytosolic, mitochondrial, and microsomal (a.k.a. MAPEG) proteins. Mammalian GSTs exist either as homodimers or heterodimers. The individual monomers were historically characterized by their quaternary structure and size, Yf (24.5kDa), Yk (25kDa), Ya1 (25.5kDa), Ya2 (25.5kDa), Yn (26.5kDa), Yb1 (27kDa), Yb2 (27kDa) and Yc (28.5kDa). The Y designation refers to the Y fraction (as opposed to the X and Z fractions) that were found during purification by Sephadex G75 chromatography. Two GST superfamilies exist that comprise either cytosolic or membrane-bound proteins. Evidence suggests that cytosolic GST increases bodily resistance to carcinogens, products of oxidative stress, environmental pollutants and certain anti-cancer drugs. The cytosolic GSTs are further subdivided into eight classes, namely alpha (GSTA), mu (GSTM), pi (GSTP), sigma (GSTS), theta (GSTT), kappa (GSTK), zeta (GSTZ), and omega (GSTO). GSTA4 exists as a homodimer composed of two Yk subunits.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GSTA4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit polyclonal antibody in serum with 0.05% sodium azide.
Gene Accession No. P14942
Gene Symbols Gsta4
Host Species Rabbit
Immunogen Purified rat liver GST K (YkYk).
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_001100310
Test Specificity This antiserum selectively reacts with Yk from purified rat liver GST K and rat kidney GST J. No reactivity toward Ya, Yc, Yb1, Yb2, Yb3, Yf, or Yn subunits from other forms of purified rat GSTs (Hayes, J.D,, and Mantle, T.J. (1986). Biochem. J. 233(3):779-788).
Target Species Mouse, Rat
Content And Storage −20°C for one year
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.