Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSTO1, Mouse, Clone: 1B6, Abnova™

Catalog No. 89002746 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-002-746 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-002-746 Supplier Abnova Corporation Supplier No. H00009446M03
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant GSTO1.

This gene encodes a member of the theta class glutathione S-transferase-like (GSTTL) protein family. In mouse, the encoded protein acts as a small stress response protein, likely involved in cellular redox homeostasis. [provided by RefSeq

Sequence: SKVPSLVGSFIRSQNKEDYAGLKEEFRKEFTKLEEVLTNKKTTFFGGNSISMIDYLIWPWFERLEAMKLNECVDHTPKLKLWMAAMKED

Specifications

Antigen GSTO1,
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1B6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant GSTO1.
Formulation PBS with no preservative; pH 7.4
Gene GSTO1
Gene Accession No. NM_004832
Gene Alias DKFZp686H13163/GSTTLp28/P28
Gene Symbols GSTO1
Host Species Mouse
Immunogen GSTO1 (NP_004823, 121 a.a. ∼ 209 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9446
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.