Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-GSTT1, Polyclonal

Catalog No. ABS1653MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABS1653MI 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS1653MI Supplier MilliporeSigma™ Supplier No. ABS1653
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects GSTT1 Clone: in Human, Mouse, Rat samples, and it is validated for Immunohistochemistry (Paraffin), Western Blotting

Glutathione S-transferase theta-1 (EC 2.5.1.18; UniProt P30711; also known as Glutathione S-transferase T1-1, GST class-theta-1, GST T1-1) is encoded by the GSTT1 gene (Gene ID 2952) in human. Glutathione S-transferases constitute a family of metabolic isozymes that are involved in detoxification by catalyzing the conjugation of the reduced form of glutathione (GSH) to xenobiotic substrates. The GST family can be subdivided into the cytosolic, mitochondrial, and microsomal (a.k.a. MAPEG) proteins. Mammalian GSTs exist as either homodimers or heterodimers. The individual monomers were historically characterized by their quaternary structure and size, Yf (24.5kDa), Yk (25kDa), Ya1 (25.5kDa), Ya2 (25.5kDa), Yn (26.5kDa), Yb1 (27kDa), Yb2 (27kDa) and Yc (28.5kDa). The Y designation refers to the Y fraction (as apposed to the X and Z fractions) they were found in during purification by Sephadex G75 chromatography. Two GST superfamilies exist that comprise either cytosolic or membrane-bound proteins. Evidence suggests that cytosolic GST increases bodily resistance to carcinogens, products of oxidative stress, environmental pollutants and certain anti-cancer drugs. The cytosolic GSTs are further subdivided into eight classes. namely alpha (GSTA), mu (GSTM), pi (GSTP), sigma, theta (GSTT), kappa (GSTK), zeta (GSTZ), and omega (GSTO). GSTT1 exists as a homodimer, the subunit is referred to as subunit 5 in rat species, and T1-1 theta subunit in human.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GSTT1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit polyclonal antibody in serum with 0.05% sodium azide.
Gene Accession No. P30711
Gene Symbols GSTT1
Host Species Rabbit
Immunogen His-tagged recombinant full-length human GSTT1.
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_000844
Test Specificity Expected to react with both spliced isoforms reported by UniProt (P30711). Note that not all human samples are positive for GSTT1 expression. It has been estimated that approx. 18% of the Caucasian, and as much as 60% of the Chinese and Korean populations are GSTT1-null (Katoh, T., et al. (1996) Carcinogenesis. 17(9):1855-1859; Lear, J.T., et al. (1996) Carcinogenesis. 17(9):1891-1896;).
Target Species Human, Mouse, Rat
Content And Storage −20°C for one year
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.