Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSTZ1, Rabbit, MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89002893 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-002-893 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-002-893 Supplier Abnova Corporation Supplier No. H00002954D01
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human GSTZ1 protein.

This gene is a member of the glutathione S-transferase (GSTs) super-family which encodes multifunctional enzymes important in the detoxification of electrophilic molecules, including carcinogens, mutagens, and several therapeutic drugs, by conjugation with glutathione. This enzyme also plays a significant role in the catabolism of phenylalanine and tyrosine. Thus defects in this enzyme may lead to severe metabolic disorders including alkaptonuria, phenylketonuria and tyrosinaemia. Several transcript variants of this gene encode multiple protein isoforms. [provided by RefSeq

Sequence: MQAGKPILYSYFRSSCSWRVRIALALKGIDYETVPINLIKDGGQQFSKDFQALNPMKQVPTLKIDGITIHQSLAIIEYLEEMRPTPRLLPQDPKKRASVRMISDLIAGGIQPLQNLSVLKQVGEEMQLTWAQNAITCGFNALEQILQSTAGIYCVGDEVTMADLCLVPQVANAERFKVDLTPYPTISSINKRLLVLEAFQVSHPCRQPDTPTELRA

Specifications

Antigen GSTZ1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human GSTZ1 protein.
Formulation No additive
Gene GSTZ1
Gene Accession No. NM_145870.1
Gene Alias GSTZ1-1/MAAI/MAI/MGC2029
Gene Symbols GSTZ1
Host Species Rabbit
Immunogen GSTZ1 (NP_665877.1, 1 a.a. ∼ 216 a.a) full-length human protein.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2954
Target Species Human, Mouse
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.