Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GT1b Ganglioside Mouse anti-Human, Mouse, Rat, Clone: GT1b-2b, MilliporeSigma™

Catalog No. MAB5608MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5608MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB5608MI Supplier MilliporeSigma Supplier No. MAB5608
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-GT1b Ganglioside Antibody is an antibody against GT1b Ganglioside for use in immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GT1b Ganglioside
Applications Immunohistochemistry
Classification Monoclonal
Clone GT1b-2b
Conjugate Unconjugated
Formulation Liquid. Contains no preservative.
Host Species Mouse
Immunogen Purified GT1b ganglioside
Purification Method Ascites
Quantity 100 μL
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.