Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-GTAP, Polyclonal

Catalog No. CB1015100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
CB1015100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CB1015100UG Supplier MilliporeSigma™ Supplier No. CB1015100UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects GTAP Clone: in Human, Mouse, Rat samples, and it is validated for Immuno Blotting

The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes (E1s), ubiquitin-conjugating enzymes (E2s), and ubiquitin-protein ligases (E3s). GTAP is a member of the E2 ubiquitin-conjugating enzyme family. GTAP may participate in intercellular adhesion and embryonic development. GTAP is thought to play a role in stem cell development via its regulation of laminin-dependent embryonic cell adhesion and the formation of embryoid bodies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GTAP
Applications Immunoblot
Classification Polyclonal
Conjugate Unconjugated
Formulation In PBS.
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 7-22 (QGQQQPGPGQQLGGQG) of human GTAP, conjugated to KLH.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽67kDa GTAP protein in HEK293 cells.
Target Species Human, Mouse, Rat
Content And Storage −20°C
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.