Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

general transcription factor IIE, polypeptide 1, alpha 56kDa, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89005485 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-005-485 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-005-485 Supplier Abnova Corporation Supplier No. H00002960B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human GTF2E1 protein.

Sequence: MADPDVLTEVPAALKRLAKYVIRGFYGIEHALALDILIRNSCVKEEDMLELLKFDRKQLRSVLNNLKGDKFIKCRMRVETAADGKTTRHNYYFINYRTLVNVVKYKLDHMRRRIETDERDSTNRASFKCPVCSSTFTDLEANQLFDPMTGTFRCTFCHTEVEEDESAMPKKDARTLLARFNEQIEPIYALLRETEDVNLAYEILEPEPTEIPALKQSKDHAATTAGAASLAGGHHREAWATKGPSYEDLYTQNVVINMDDQEDLHRASLEGKSAKERPIWLRESTVQGAYGSEDMKEGGIDMDAFQEREEGHAGPDDNEEVMRALLIHEKKTSSAMAGSVGAAAPVTAANGSDSESETSESDDDSPPRPAAVAVHKREEDEEEDDEFEEVADDPIVMVAGRPFSYSEVSQRPELVAQMTPEEKEAYIAMGQRMFEDLFE

Specifications

Antigen general transcription factor IIE, polypeptide 1, alpha 56kDa
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human GTF2E1 protein.
Formulation No additive
Gene GTF2E1
Gene Accession No. NM_005513.1
Gene Alias FE/TF2E1/TFIIE-A
Gene Symbols GTF2E1
Host Species Mouse
Immunogen GTF2E1 (NP_005504.2, 1 a.a. ∼ 439 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Stem Cell Biology
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 2960
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.