Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
general transcription factor IIIC, polypeptide 3, 102kDa, Mouse, Clone: 3D9, Abnova™
Mouse monoclonal antibody raised against a partial recombinant GTF3C3.
Supplier: Abnova Corporation H00009330M02
Description
Sequence: TPEQPTAGDVFVLEMVLNRETKKMMKEKRPRSKLPRALRGLMGEANIRFARGEREEAILMCMEIIRQAPLAYEPFSTLAMIYEDQGDMEKSLQFELIAAHLNPSpecifications
general transcription factor IIIC, polypeptide 3, 102kDa | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_012086 | |
GTF3C3 | |
GTF3C3 (NP_036218, 112 a.a. ∼ 214 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Yes | |
9330 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG2b κ |
ELISA, KnockDown, Western Blot | |
3D9 | |
Mouse monoclonal antibody raised against a partial recombinant GTF3C3. | |
GTF3C3 | |
TFIIIC102/TFIIICgamma/TFiiiC2-102 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction