Learn More
GTP binding protein 1 (A01), Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant GTPBP1.
Supplier: Abnova Corporation H00009567A01
Description
This gene is upregulated by interferon-gamma and encodes a protein that is a member of the AGP11/GTPBP1 family of GTP-binding proteins. A structurally similar protein has been found in mouse, where disruption of the gene for that protein had no observable phenotype. [provided by RefSeq
Sequence: MDEGCGETIYVIGQGSDGTEYGLSEADMEASYATVKSMAEQIEADVILLRERQEAGGRVRDYLVRKRVGDNDFLEVSpecifications
GTP binding protein 1 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant GTPBP1. | |
GTPBP1 | |
GP-1/GP1/HSPC018/MGC20069 | |
Mouse | |
50 μL | |
Yes | |
9567 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_004286 | |
GTPBP1 | |
GTPBP1 (NP_004277, 1 a.a. ∼ 76 a.a) partial recombinant protein with GST tag. | |
RUO | |
Primary | |
Human | |
Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.