Learn More
GTP binding protein 1, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Shop All Abnova Corporation ProductsDescription
This gene is upregulated by interferon-gamma and encodes a protein that is a member of the AGP11/GTPBP1 family of GTP-binding proteins. A structurally similar protein has been found in mouse, where disruption of the gene for that protein had no observable phenotype. [provided by RefSeq
Specifications
Specifications
| Antigen | GTP binding protein 1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Rabbit polyclonal antibody raised against a full-length human GTPBP1 protein. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | GTPBP1 |
| Gene Accession No. | ENST00000335466 |
| Gene Alias | GP-1/GP1/HSPC018/MGC20069 |
| Gene Symbols | GTPBP1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.