Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GUCA1A, Mouse, Clone: 2F7, Abnova™

Catalog No. 89022429 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-022-429 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-022-429 Supplier Abnova Corporation Supplier No. H00002978M04
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant GUCA1A.

This gene plays a role in the recovery of retinal photoreceptors from photobleaching. In the recovery phase, the phototransduction messeneger cGMP is replenished by retinal guanylyl cyclase-1 (GC1). GC1 is activated by decreasing Ca(2+) concentrations following photobleaching. The protein encoded by this gene, guanylyl cyclase activating protein 1 (GCAP1), mediates the sensitivity of GC1 to Ca(2+) concentrations. GCAP1 promotes activity of GC1 at low Ca(2+) concentrations and inhibits GC1 activity at high Ca(2+) concentrations. Mutations in this gene cause autosomal dominant cone dystrophy (COD3); a disease characterized by reduced visual acuity associated with progressive loss of color vision. Mutations in this gene prohibit the inactivation of RetGC1 at high Ca(2+) concentrations; causing the constitutive activation of RetGC1 and, presumably, increased cell death. This gene is expressed in retina and spermatagonia. [provided by RefSeq]

Sequence: MGNVMEGKSVEELSSTECHQWYKKFMTECPSGQLTLYEFRQFFGLKNLSPSASQYVEQMFETFDFNKDGYIDFMEYVAALSLVLKGKVEQKLR

Specifications

Antigen GUCA1A
Applications ELISA, KnockDown, Western Blot
Classification Monoclonal
Clone 2F7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant GUCA1A.
Formulation PBS with no preservative; pH 7.4
Gene GUCA1A
Gene Accession No. NM_000409
Gene Alias COD3/GCAP/GCAP1/GUCA/GUCA1
Gene Symbols GUCA1A
Host Species Mouse
Immunogen GUCA1A (NP_000400, 1 a.a. ∼ 93 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2978
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.