Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GUCA1B, Mouse, Clone: 5E7, Abnova™

Catalog No. 89022430 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-022-430 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-022-430 Supplier Abnova Corporation Supplier No. H00002979M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant GUCA1B.

Sequence: KLKWTFKIYDKDGNGCIDRLELLNIVEGIYQLKKACRRELQTEQDQLLTPEEVVDRIFLLVDENGDGQLSLNEFVEGARRDKWVMKMLQMDMNPSSWLAQQRRKSAMF

Specifications

Antigen GUCA1B
Applications ELISA, Western Blot
Classification Monoclonal
Clone 5E7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant GUCA1B.
Formulation PBS with no preservative; pH 7.4
Gene GUCA1B
Gene Accession No. NM_002098
Gene Alias DKFZp686E1183/GCAP2/GUCA2
Gene Symbols GUCA1B
Host Species Mouse
Immunogen GUCA1B (NP_002089, 93 a.a. ∼ 200 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2979
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.