Learn More
guanylate cyclase 1, soluble, alpha 2, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant GUCY1A2.
Supplier: Abnova Corporation H00002977A01
Description
Soluble guanylyl (or guanylate) cyclases are heterodimeric enzymes consisting of an alpha subunit, such as alpha-2 (GUCY1A2), and a beta subunit, typically beta-1 (GUCY1B3; MIM 139397), which are activated by nitric oxide (NO) and which catalyze conversion of GTP to 3-prime, 5-prime-cyclic GMP and pyrophosphate (Harteneck et al., 1991 [PubMed 1683630]).[supplied by OMIM
Sequence: CENTNIMKNLPQGTSQVPADLRISINTFCRAFPFHLMFDPSMSVLQLGEGLRKQLRCDTHKVLKFEDCFEIVSPKVNATFERVLLRLSTPFVIRTKPEASGSENKDSpecifications
guanylate cyclase 1, soluble, alpha 2 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant GUCY1A2. | |
GUCY1A2 | |
GC-SA2/GUC1A2 | |
Mouse | |
50 μL | |
Signal Transduction | |
Primary | |
Human | |
Serum |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_000855 | |
GUCY1A2 | |
GUCY1A2 (NP_000846, 296 a.a. ∼ 401 a.a) partial recombinant protein with GST tag. | |
RUO | |
Yes | |
2977 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.