Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GUCY1A3, Mouse, Clone: 2H1, Abnova™

Catalog No. 89022431 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-022-431 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-022-431 Supplier Abnova Corporation Supplier No. H00002982M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant GUCY1A3.

Soluble guanylate cyclase (sGC), a heterodimeric protein consisting of an alpha subunit, such as alpha-1 (GUCY1A3), and a beta subunit, typically beta-1 (GUCY1B3; MIM 139397), catalyzes conversion of GTP to the second messenger cGMP and functions as the main receptor for nitric oxide and nitrovasodilator drugs (Zabel et al., 1998 [PubMed 9742212]).[supplied by OMIM

Sequence: KATMPICQDIPEKNIQESLPQRKTSRSRVYLHTLAESICKLIFPEFERLNVALQRTLAKHKIKESRKSLEREDFEKTIAEQAVAAGVPVEVIKESLGEEV

Specifications

Antigen GUCY1A3
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2H1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant GUCY1A3.
Formulation PBS with no preservative; pH 7.4
Gene GUCY1A3
Gene Accession No. BC028384
Gene Alias GC-SA3/GUC1A3/GUCA3/GUCSA3/GUCY1A1
Gene Symbols GUCY1A3
Host Species Mouse
Immunogen GUCY1A3 (AAH28384, 41 a.a. ∼ 140 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2982
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.