Learn More
GYPA, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human GYPA peptide using ARM Technology.
Supplier: Abnova Corporation H00002993K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
Glycophorins A (GYPA) and B (GYPB) are major sialoglycoproteins of the human erythrocyte membrane which bear the antigenic determinants for the MN and Ss blood groups. In addition to the M or N and S or s antigens that commonly occur in all populations, about 40 related variant phenotypes have been identified. These variants include all the variants of the Miltenberger complex and several isoforms of Sta, as well as Dantu, Sat, He, Mg, and deletion variants Ena, S-s-U- and Mk. Most of the variants are the result of gene recombinations between GYPA and GYPB. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
| GYPA | |
| Monoclonal | |
| Unconjugated | |
| In 1X PBS, pH 7.4 | |
| GYPA | |
| GYPA | |
| A synthetic peptide of human GYPA | |
| RUO | |
| 2993 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| ELISA, Western Blot | |
| 1mg | |
| glycophorin A (MNS blood group) | |
| GYPA | |
| CD235a, GPA, GPErik, GPSAT, GpMiIII, HGpMiIII, HGpMiV, HGpMiX, HGpMiXI, HGpSta(C), MN, MNS | |
| Rabbit | |
| 1 mg x up to 3 Abs | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.