Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H-2D[b] Mouse anti-Mouse, FITC, Clone: KH95, BD

Catalog No. BDB553573 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB553573 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB553573 Supplier BD Biosciences Supplier No. 553573
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The KH95 antibody reacts with the H-2D[b] MHC class I alloantigen. Reactivity with other haplotypes (e.g., a, d, f, k, n, p, q, r, s, u, v) has not been observed.

Host Species: Mouse
Clone: KH95
Isotype: IgG2b, κ
Species Reactivity: Mouse
Immunogen: C57BL/10 mouse skin graft and splenocytes

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen H-2D[b]
Applications Flow Cytometry
Classification Monoclonal
Clone KH95
Concentration 0.5mg/mL
Conjugate FITC
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen C57BL/10 mouse skin graft and splenocytes
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.