Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H-2D[d] Mouse anti-Mouse, Unlabeled, Clone: 34-2-12, BD

Catalog No. BDB553577 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB553577 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB553577 Supplier BD Biosciences Supplier No. 553577
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 34-2-12 antibody (also known as 34-2-12S) recognizes the α3 domain of the H-2D[d]. The binding of the antibody to its epitope isindependent of the α1 and α2 domains and β2 microglobulin. It cross-reacts with cells of the C3H.LG/Ckc strain. Reactivity with otherhaplotypes (eg, b, f, k, p, q, r, s) has not been observed. Soluble mAb 34-2-12 blocks binding of the Ly-49A-expressing T lymphoma EL4 toimmobilized H-2D[d]. However, further studies utilizing this mAb indicate that the α3 domain is not involved in the interaction betweenLy-49A, or Ly-49G2, and H-2D[d]. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Mouse
Clone: 34-2-12
Isotype: IgG2a κ
Species Reactivity: Mouse
Immunogen: (C57BL/6 x DBA/2)F1 mouse splenocytes

Flow Cytometry, Immunoprecipitation

TRUSTED_SUSTAINABILITY

Specifications

Antigen H-2D[d]
Applications Flow Cytometry
Classification Monoclonal
Clone 34-2-12
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen (C57BL/6 x DBA/2)F1 mouse splenocytes
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.