Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H-2D(d) Mouse anti-Mouse, Unlabeled, Clone: 34-5-8S, BD

Catalog No. BDB558915 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558915 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558915 Supplier BD Biosciences Supplier No. 558915

Mouse Monoclonal Antibody

The 34-5-8S antibody recognizes an epitope on the N-terminal domains, α1 and α2, of the H-2Dd. The mAb identifies a conformationally sensitive epitope of H-2Dd associated with β2 microglobulin; it fails to react with free H-2D(d) α chains synthesized in vitro. Weak cross-reactivity with cells from mice of the H-2b, H-2q, and H-2s haplotypes has been observed by flow cytometric analysis. Reactivity with other haplotypes (e.g., f, k, p, r) has not been observed. mAb 34-5-8S has been reported to block the recognition of H-2Dd by Ly-49A+, C+, F+, G2+, or I+ NK cells or transfectants. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Flow Cytometry, Immunoprecipitation

TRUSTED_SUSTAINABILITY

Specifications

Antigen H-2D(d)
Applications Flow Cytometry
Classification Monoclonal
Clone 34-5-8S
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen (C57BL/6 x DBA/2)F1 hybrid mouse splenocytes
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.