Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
H-2Kd/H-2Dd Mouse anti-Mouse, Biotin, Clone: 34-1-2S, Southern Biotech™

Mouse Monoclonal Antibody
Supplier: Southern Biotechnology Associates 191108
Description
SouthernBiotech offers a variety of anti-mouse primary antibodies which are available in numerous formats including Alexa Fluor and Pacific Blue.
Specifications
| H-2Kd/H-2Dd | |
| Monoclonal | |
| 0.5mg/mL | |
| Supplied in PBS/NaN3 | |
| BDF1 mouse splenocytes | |
| RUO | |
| Mouse | |
| IgG2a |
| Flow Cytometry, Immunocytochemistry, Immunoprecipitation | |
| 34-1-2S | |
| Biotin | |
| Mouse | |
| 0.5 mg | |
| Primary | |
| Store at 2-8°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction