Learn More
H-2K[b]/H-2D[b] Mouse anti-Mouse, Unlabeled, Clone: 28-8-6, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 553575
Description
The 28-8-6 antibody reacts with the H-2K[b] and H-2D[b] MHC class I alloantigens. The reaction with H-2K[b] has been reported to be stronger than that with H-2D[b]. Its recognition of H-2D[b] is dependent upon the presence of β2 microglobulin. Cross-reactivity with an epitope on the N-terminal domains, α1 and α2, of the H-2D[d] MHC class I alloantigen has also been described. Reactivity with other haplotypes (e.g., f, k, p, q, r, s) has not been observed.
Host Species: Mouse
Clone: 28-8-6
Isotype: IgG2a κ
Species Reactivity: Mouse
Immunogen: C3H.SW mouse splenocytes
Flow Cytometry
Specifications
| H-2K[b]/H-2D[b] | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| C3H.SW mouse splenocytes | |
| 0.5 mg | |
| Immunology | |
| Mouse | |
| IgG2a κ |
| Flow Cytometry | |
| 28-8-6 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.