Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H-CAM Mouse anti-Human, Clone: F10-44-2, MilliporeSigma™

Catalog No. CBL154MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CBL154MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CBL154MI Supplier MilliporeSigma Supplier No. CBL154
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-H-CAM Antibody, clone F10-44-2 is validated for use in flow cytometry, immunoprecipitation, immunohistochemistry, immunohistochemistry (paraffin) for the detection of H-CAM.
TRUSTED_SUSTAINABILITY

Specifications

Antigen H-CAM
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone F10-44-2
Conjugate Unconjugated
Formulation The monoclonal is presented at a concentration of 100μg/mL in phosphate buffered saline containing 10mM sodium azide and 1mg/ml bovine serum albumin. We recommend that each laboratory determine an optimum working titre for use in its particular applic
Gene Accession No. NM_000610.3; NM_001001389.1; NM_001001390.1; NM_001001391.1; NM_001001392.1
Gene Symbols CD44, LHR, HUTCH-I, CDW44, CDw44, ECMR-III, Epican, MGC10468, CD44R, MDU2, PGP-1, CSPG8, Pgp1, IN, MUTCH-I, MDU3, MIC4, MC56, HCELL
Host Species Mouse
Immunogen Purified human T cells
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 4°C for up to 1 month, −20°C in undiluted aliquots for long term
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.